missing translation for 'onlineSavingsMsg'
Learn More

beta Tubulin Loading Control Monoclonal Antibody (BT7R), DyLight™ 800 4X PEG, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA516308D800

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15511164

  • £350.00 / 50µL
Estimated Shipment: 12-08-2024
to see stock.

Description

Description

MA5-16308-D800 detects bet-Tubulin from various species. MA5-16308-D800 has been successfully used for fluorescent detection of beta-Tubulin in Western Blotting. DyLight 800 4X PEG has an excitation/emission of 783/797 nm.

beta Tubulin Loading Control Monoclonal antibody specifically detects beta Tubulin Loading Control in Human, Mouse, Non-human Primate, Rat samples. It is validated for Western Blot.
Specifications

Specifications

beta Tubulin Loading Control
Monoclonal
1 mg/mL
MES with proprietary stabilizer and no preservative
A2AQ07, P04350, P07437, P69897, P85108, P99024, Q13509, Q13885, Q3ZCM7, Q4QRB4, Q7TMM9, Q922F4, Q9BUF5, Q9D6F9, Q9ERD7, Q9H4B7
TUBB, Tubb1, Tubb2a, Tubb3, Tubb4a, Tubb5, Tubb6, TUBB8
KLH conjugated peptide of the N-terminus of Beta-Tubulin.
50 μL
Primary
Human, Mouse, Non-human Primate, Rat
Antibody
IgG2a
Western Blot
BT7R
DyLight 800 4X PEG
TUBB8
2310057H16Rik; 2810484G07Rik; 3200002H15Rik; AA408537; AI325297; AI596182; B130022C14Rik; bA631M21.2; BB220206; beta 5-tubulin; beta Ib tubulin; beta tubulin; beta tubulin 1; beta-4; beta-4 tubulin; beta-4-tubulin; beta-5; beta-5 tubulin; beta5-tubulin; betaIII-tubulin; beta-tubulin; beta-tubulin class-III; Beta-tubulin class-V; beta-tubulin class-VI; Beta-tubulin isotype I; c(beta)7 tubulin; CDCBM; CDCBM1; CDCBM5; CDCBM6; CFEOM3; CFEOM3A; class I beta tubulin; Class I beta-tubulin; class IIa beta-tubulin; class III beta-tubulin; class IVa beta-tubulin; class V beta tubulin; class V beta-tubulin; CSCSC1; dJ40E16.7; dystonia 4, torsion (autosomal dominant); DYT4; FEOM3; HSA10p15 beta-tubulin 4Q; HsT1601; hypothetical protein LOC540236; hypothetical protein LOC550458; I79_000194; I79_012097; LOC100349792; LOC100761889; LOW QUALITY PROTEIN: tubulin beta-6 chain; M(beta)1; M(beta)2; M(beta)3; M(beta)4; M(beta)5; M(beta)6; M40; M40 antibody; MGC128415 protein; MGC137802 protein; neuron-spec
Mouse
Protein A
RUO
10381, 10382, 203068, 22151, 22152, 22153, 22154, 246118, 29213, 29214, 307351, 347688, 498736, 545486, 679312, 67951, 7280, 81027, 84617
4° C, do not freeze
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers