missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-BKV VP1, Clone: 3G2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB3204-M23.50ug
Additional Details : Weight : 0.00010kg
Specifications
BKV VP1 | |
Monoclonal | |
Unconjugated | |
In 1x PBS, pH 7.4 | |
VP1 | |
Mouse | |
50 μg | |
Primary | |
Virus | |
Purified |
ELISA, Western Blot | |
3G2 | |
Mouse monoclonal antibody raised against a full-length recombinant BKV VP1. | |
NC_001538 | |
BKPyVgp4 | |
BKV VP1 (YP_717939.1, 1 a.a. ∼ 369 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
1489515 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
For Research Use Only