missing translation for 'onlineSavingsMsg'
Learn More

anti-BST1 Matched Antibody Pair, Abnova™

Brand:  Abnova H00000683-AP11.1Set

Additional Details : Weight : 0.00010kg

Product Code. 16101794

  • £660.00 / 1 set
Estimated Shipment: 21-08-2024
to see stock.

Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-BST1 (100μg); Detection antibody: mouse monoclonal anti-BST1, IgG2a κ, (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols

Description

Description

This antibody pair set comes with matched antibody pair to detect and quantify protein level of human BST1

Host Species: Rabbit, Mouse
Isotype: IgG, IgG2a κ
Species Reactivity: Hu

ELISA Pair (Recombinant Protein)

Specifications

Specifications

BST1
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human BST1.
CD157
Mouse, Rabbit
1 Set
Primary
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-BST1 (100 μg) 2. Detection antibody: mouse monoclonal anti-BST1, IgG2a Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
ELISA
Unconjugated
BST1
BST1
Affinity Purified
RUO
683
Human
IgG, IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers