missing translation for 'onlineSavingsMsg'
Learn More

CAPZB, Mouse, Clone: 1H1, Abnova™

Mouse monoclonal antibody raised against a partial recombinant CAPZB.

Brand:  Abnova H00000832-M02.100ug

Product Code. 16105644

  • £283.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

CAPZB is a member of the F-actin capping protein family. This gene encodes the beta subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments. [provided by RefSeq

Sequence: SLTRQMEKDETVSDCSPHIANIGRLVEDMENKIRSTLNEIYFGKTKDIVNGLRSVQTFADKSKQEALKNDLVEALKRKQQC
Specifications

Specifications

CAPZB
Monoclonal
Unconjugated
PBS with no preservative; pH 7.4
NM_004930
CAPZB
CAPZB (NP_004921, 192 a.a. ∼ 272 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA, Immunohistochemistry (PFA fixed), Western Blot
1H1
Mouse monoclonal antibody raised against a partial recombinant CAPZB.
CAPZB
CAPB/CAPPB/CAPZ/MGC104401/MGC129749/MGC129750
Mouse
Affinity chromatography
RUO
832
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.