missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD36 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527236
This item is not returnable.
View return policy
Description
Recommended positive controls: K562, mouse brown adipose, RAW264.7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CD36, also known as scavenger receptor class B member 3, is a protein that is expressed on the surface of various cell types, including macrophages, platelets, and adipocytes. It plays a role in lipid metabolism, inflammation, and atherosclerosis, and is involved in the recognition and uptake of various ligands such as oxidized low-density lipoproteins, long-chain fatty acids, and apoptotic cells. CD36 is also implicated in the pathogenesis of malaria. The protein encoded by this gene serves as a receptor for thrombospondin in platelets and various cell lines, and is the fourth major glycoprotein of the platelet surface. It binds to collagen, thrombospondin, anionic phospholipids, and oxidized LDL, and directly mediates cytoadherence of Plasmodium falciparum parasitized erythrocytes. Mutations in this gene cause platelet glycoprotein deficiency. Multiple alternatively spliced transcript variants have been found for this gene. Diseases associated with CD36 include Platelet Glycoprotein IV Deficiency and Coronary Heart Disease 7.Specifications
CD36 | |
Polyclonal | |
Unconjugated | |
CD36 | |
adipocyte membrane protein; BDPLT10; Cd36; CD36 antigen; CD36 antigen (collagen type I receptor, thrombospondin receptor); CD36 molecule; CD36 molecule (thrombospondin receptor); CD36 protein; CHDS7; cluster determinant 36; collagen type I receptor thrombospondin receptor; collagen type I receptor, thrombospondin receptor; FAT; FAT/CD36; fatty acid translocase; fatty acid translocase/CD36; fatty acid transport protein; glycoprotein IIIb; GP3B; GP4; GPIIIB; GPIV; I79_011940; Leukocyte differentiation antigen CD36; PAS IV; PAS4; PAS-4; PAS-4 protein; PASIV; Platelet collagen receptor; Platelet glycoprotein 4; platelet glycoprotein IV; Scarb3; scavenger receptor class B, member 3; SR-B3; Thrombospondin receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12491, 948 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.47 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
P16671, Q08857 | |
CD36 | |
Synthetic peptide corresponding to a region within amino acids 302 and 395 of Human CD36 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |