Learn More
CD4 Mouse anti-Human, BD IMag-DM, Clone: L200, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 557767
Additional Details : Weight : 0.02000kg
Description
BD IMag™ anti-human CD4 Particles - DM are magnetic nanoparticles that have monoclonal antibody conjugated to their surfaces. These particles are optimized for the positive selection or depletion of CD4-bearing T lymphocytes using the BD IMagnet™. The L200 antibody reacts with CD4 on human, rhesus and cynomolgus macaque, and baboon peripheral blood leukocytes; we have confirmed that the BD IMag™ particles can effectively separate the CD4-bearing cells of rhesus macaque blood. The distribution of CD4 on peripheral leukocytes is similar for both human and monkey. It is on the MHC class II-restricted T helper cells, with the majority of CD4-positive lymphocytes being CD8-negative. It is also found on most thymocytes and at low density on monocytes; it is not found on B or NK cells.
Specifications
CD4 | |
Monoclonal | |
BD IMag-DM | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human HPB-ALL Cell Line | |
1 x 109 Cells | |
Immunology | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
IgG1 κ |
Flow Cytometry | |
L200 | |
L3T4 ; T-cell surface antigen T4/Leu-3; W3/25 ; CD4 antigen (p55) | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
For Research Use Only.