missing translation for 'onlineSavingsMsg'
Learn More

CD8a Rat anti-Mouse, BD IMag-DM, Clone: 53-6.7, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 551516

Additional Details : Weight : 0.02000kg

Product Code. 15856678

  • £1305.00 / Each
Estimated Shipment: 22-08-2024
to see stock.

Description

Description

BD IMag™ anti-mouse CD8a Particles - DM are magnetic nanoparticles that have monoclonal antibody conjugated to their surfaces. These particles are optimized for the positive selection or depletion of CD8a-bearing leukocytes using the BD IMagnet™. CD8a has been reported to be expressed on most thymocytes and a subpopulation of mature T lymphocytes (e.g. MHC class I-restricted T cells, including most T suppressor/cytotoxic cells). In addition, subsets of γδ TCR-bearing T cells, intestinal intraepithelial lymphocytes, and dendritic cells also have been reported to express CD8a. Leukocytes are labeled with BD IMag™ anti-mouse CD8a Particles - DM according to the Magnetic Labeling Protocol. This labeled cell suspension is then placed within the magnetc field of BD IMagnet™ (Cat. No. 552311). Labeled cells migrate toward the magnet (positive fraction), leaving the unlabeled cells in suspension so they can be drawn off (negative fraction). The tube is then removed from the magnetic field for resuspension of the positive fraction. The seperation is repeated twice to increase the purity of the positive fraction. The magnetic separation steps are diagrammed in the Separation Flow Chart. After the positive fraction is washed, the small size of the magnetic particles allows the positive fraction to be further evaluated in downstream applications such as flow cytometry.

Host Species:
Clone: 53-6.7

Cell Separation

Specifications

Specifications

CD8a
Monoclonal
BD IMag-DM
Rat
10 mL
Immunology
Mouse
IgG2a κ
Flow Cytometry
53-6.7
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse Spleen Cells or Thymocyte Membranes
RUO
Primary
Store undiluted at 4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.