Learn More
anti-CDC42, MAPK8 Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0572.1Set
Additional Details : Weight : 0.00010kg
Includes: CDC42 rabbit purified polyclonal antibody (20μg); MAPK8 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the CDC42 protein, and the other against the MAPK8 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
CDC42 | |
Monoclonal/Polyclonal | |
CDC42 | |
CDC42 | |
1 Set | |
Primary | |
Antibody pair set content: 1. CDC42 rabbit purified polyclonal antibody (20 ug) 2. MAPK8 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
CDC42Hs/G25K | |
Mouse, Rabbit | |
RUO | |
998/5599 | |
Human | |
CDC42 & MAPK8 Protein Protein Interaction Antibody Pair |