missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDR2, Mouse, Clone: 4F5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CDR2.
Brand: Abnova H00001039-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: LTVPESHRKPLKRSSSETILSSLAGSDIVKGHEETCIRRAKAVKQRGISLLHEVDTQYSALKVKYEELLKKCQEEQDSLSHKAVQTSRAAAKDLTGVNAQSEPVASGWESpecifications
CDR2 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_001802 | |
CDR2 | |
CDR2 (NP_001793, 296 a.a. ∼ 404 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunofluorescence, Western Blot | |
4F5 | |
Mouse monoclonal antibody raised against a partial recombinant CDR2. | |
CDR2 | |
CDR62/Yo | |
Mouse | |
Affinity chromatography | |
RUO | |
1039 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction