missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Anti-Clumping Agent
GREENER_CHOICE

Product Code. 15234475
Click to view available options
Quantity:
20 mL
100 mL
Unit Size:
100mL
20mL
This item is not returnable. View return policy

Product Code. 15234475

Brand: Gibco™ 0010057DG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Anti-Clumping Agent

GIBCO™ Anti-Clumping Agent is a concentrated liquid reagent that reduces cell clumping to assist in attaining higher viable cell densities in suspension culture. Anti-Clumping Agent is a gentle, animal origin-free, chemically defined formulation that contains no proteins, enzymes, hydrolysates, or components of unknown composition.

  • Reduced cell clumping in suspension culture
  • Higher viable cell densities
  • Animal origin-free and chemically defined components
  • Convenient packaging with a larger size now available
  • Customization available to configure packaging and delivery

    Simple and Gentle Cell Dissociation
    Cell lines adapted to suspension culture can revert to a more adherent or aggregated clumping state. When this occurs, culture condition can be greatly improved by adding a gentle reagent to reduce clumping and adhesion instead of the harsh enzymes often found in cell dissociation products. Anti-Clumping Agent is a simple, gentle, concentrated solution which, when added to culture media (usually titrated between a 1:100 and 1:1000 dilution), can reduce cell clumping significantly.

    Reduced Clumping, Increased Viability, Improved Protein Expression
    Reduced cell clumping achieved through use of Anti-Clumping Agent is illustrated below (Fig 1). See also the data demonstrating improved viable cell density (Fig 2). Higher viable cell density contributes to the stabilization of culture conditions for better protein yield.

    For actual cell dissociation, we recommend our animal origin-free TrypLE™ products.

    Use Anti-Clumping Agent after Transfection
    Anti-Clumping Agent is not recommended in all suspension cell culture conditions. For example, when creating a cell line using lipid cationic transfection, Anti-Clumping Agent can interfere with the transfection process, so in this case we recommend that Anti-Clumping Agent be used after transfection.

    Animal Origin-free and Chemically Defined Components
    Because Anti-Clumping Agent is made with only animal origin free and chemically defined components, you can be assured that the risk of adding adventitious agents to your culture system is minimized. Avoiding additives in your system that are not well defined can lead to more robust results from batch to batch.

    Convenient and Customizable Packaging
    Previously, Anti-Clumping Agent was only available in a 20 mL bottle. However, it is now available in a more economical and convenient 100 mL bottle. For even larger packaging, you can custom configure this product by clicking on the 'Customize this Product' link under the 'Add to Cart' button above or by emailing us at CustomMedia@lifetech.com.

    Additional Information and Support
    Please review the Manuals and Protocols provided below for additional information on successful use of this product. Direct any unanswered questions to our Technical Services group.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2 to 8°C
Cell Line CHO, Insect Cells, 293 (HEK)
Cell Type Mammalian Cells
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture Type Suspension Cell Culture
Form Liquid
Product Type Anti-clumping Reagent
Format Bottle(s)
pH 6 to 8
Product Line Gibco
Quantity 100 mL
Shipping Condition Room Temperature
Source Animal origin-free
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.