missing translation for 'onlineSavingsMsg'
Learn More

anti-FAS, DAXX Protein Protein Interaction Antibody Pair, Abnova™

Brand:  Abnova DI0523.1Set

Additional Details : Weight : 0.00010kg

Product Code. 16141434

  • £529.00 / 1 set
Estimated Shipment: 06-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: FAS rabbit purified polyclonal antibody (20μg); DAXX mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols

Description

Description

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the FAS protein, and the other against the DAXX protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Specifications

FAS
Monoclonal/Polyclonal
FAS
FAS
1 Set
Primary
Antibody pair set content: 1. FAS rabbit purified polyclonal antibody (20 ug) 2. DAXX mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Purified
In situ PLA
Unconjugated
ALPS1A/APO-1/APT1/CD95/FAS1/FASTM/TNFRSF6
Mouse, Rabbit
RUO
355/1616
Human
FAS & DAXX Protein Protein Interaction Antibody Pair
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers