missing translation for 'onlineSavingsMsg'
Learn More

anti-FASLG, PIK3R1 Protein Protein Interaction Antibody Pair, Abnova™

Brand:  Abnova DI0455.1Set

Additional Details : Weight : 0.00010kg

Product Code. 16101384

  • £529.00 / 1 set
Estimated Shipment: 06-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: FASLG rabbit purified polyclonal antibody (20μg); PIK3R1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols

Description

Description

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the FASLG protein, and the other against the PIK3R1 protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Specifications

FASLG
Monoclonal/Polyclonal
FASLG
FASLG
1 Set
Primary
Antibody pair set content: 1. FASLG rabbit purified polyclonal antibody (20 ug) 2. PIK3R1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Purified
In situ PLA
Unconjugated
APT1LG1/CD178/CD95L/FASL/TNFSF6
Mouse, Rabbit
RUO
356/5295
Human
FASLG & PIK3R1 Protein Protein Interaction Antibody Pair
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers