missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitritex™ Anti-Fog Spray
Address any goggle fogging issues with an application of this spray. Nitritex™ Anti-Fog Spray is quick and easy to apply.
Brand: Nitritex™ BAFS-601
8.05 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Additional Details : Weight : 0.07200kg
Description
- Reduces fogging
- Airless pump action spray
- Non-hazardous
- Colorless
- Gamma-irradiated
- Alcohol free
- Quick and easy application
- Suitable for all BioClean autoclavable goggles
Specifications
Manufactured in a facility holding ISO 9001:2008 certifications | |
Goggles | |
60 mL |
Anti-fog spray | |
Anti-fogging applications |