missing translation for 'onlineSavingsMsg'
Learn More

glutamate-ammonia ligase (glutamine synthetase), Mouse, Clone: 3B6, Abnova™

Mouse monoclonal antibody raised against a partial recombinant GLUL.

Brand:  Abnova H00002752-M02.100ug

Additional Details : Weight : 0.00010kg

Product Code. 16118754

  • £273.00 / 100µg
Estimated Shipment: 12-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Glutamine is a main source of energy and is involved in cell proliferation, inhibition of apoptosis, and cell signaling (Haberle et al., 2005 [PubMed 16267323]). Fetal glutamine requirements are very high and depend largely on active glutamine synthesis and the release of glutamine into the fetal circulation by the placenta. Glutamine synthetase (EC 6.3.1.2), also called glutamate-ammonia ligase (GLUL), is expressed throughout the body and plays an important role in controlling body pH and in removing ammonia from the circulation. The enzyme clears L-glutamate, the major neurotransmitter in the central nervous system, from neuronal synapses (see references in Clancy et al., 1996 [PubMed 8975719]).[supplied by OMIM

Sequence: IEKLSKRHQYHIRAYDPKGGLDNARRLTGFHETSNINDFSAGVANRSASIRIPRTVGQEKKGYFEDRRPSANCDPFSVTEALIRTCLLNETGDEPFQYKN
Specifications

Specifications

glutamate-ammonia ligase (glutamine synthetase)
Monoclonal
Unconjugated
PBS with no preservative; pH 7.4
NM_002065
GLUL
GLUL (NP_002056, 274 a.a. ∼ 373 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Yes
2752
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG1 κ
ELISA, Western Blot
3B6
Mouse monoclonal antibody raised against a partial recombinant GLUL.
GLUL
GLNS/GS/PIG43/PIG59
Mouse
Affinity Purified
RUO
Primary
Human, Mouse, Rat
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers