Learn More
anti-GST, Polyclonal, Cytiva
Detect recombinant schistosomal GST fusion proteins expressed using pGEX vectors. Anti-GST, Polyclonal, Cytiva, is optimized for sensitive and specific applications.
Brand: Life Science products Cytiva 27-4577-01
Code : GE
Additional Details : Weight : 0.03100kg
Description
Host Species: Goat
Immunogen: Purified schistosomal GST
Dot Blotting, Western Blotting
Specifications
GST | |
Goat | |
Affinity Purified | |
Primary |
Unconjugated | |
Purified schistosomal GST | |
500 μL (50 Detections) |