Learn More
H-2D[d] Mouse anti-Mouse, Unlabeled, Clone: 34-2-12, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 553577
Description
The 34-2-12 antibody (also known as 34-2-12S) recognizes the α3 domain of the H-2D[d]. The binding of the antibody to its epitope isindependent of the α1 and α2 domains and β2 microglobulin. It cross-reacts with cells of the C3H.LG/Ckc strain. Reactivity with otherhaplotypes (eg, b, f, k, p, q, r, s) has not been observed. Soluble mAb 34-2-12 blocks binding of the Ly-49A-expressing T lymphoma EL4 toimmobilized H-2D[d]. However, further studies utilizing this mAb indicate that the α3 domain is not involved in the interaction betweenLy-49A, or Ly-49G2, and H-2D[d]. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: 34-2-12
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: (C57BL/6 x DBA/2)F1 mouse splenocytes
Flow Cytometry, Immunoprecipitation
Specifications
| H-2D[d] | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| (C57BL/6 x DBA/2)F1 mouse splenocytes | |
| 0.5 mg | |
| Immunology | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| 34-2-12 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.