missing translation for 'onlineSavingsMsg'
Learn More

histidine ammonia-lyase, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against a full-length human HAL protein.

Brand:  Abnova H00003034-D01P.100ug

Additional Details : Weight : 0.00010kg

Product Code. 16118647

  • £286.00 / 100µg
Estimated Shipment: 12-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Histidine ammonia-lyase is a cytosolic enzyme catalyzing the first reaction in histidine catabolism, the nonoxidative deamination of L-histidine to trans-urocanic acid. Histidine ammonia-lyase defects cause histidinemia which is characterized by increased histidine and histamine and decreased urocanic acid in body fluids [provided by RefSeq

Sequence: MPRYTVHVRGEWLAVPCQDAQLTVGWLGREAVRRYIKNKPDNGGFTSVDDAHFLVRRCKGLGLLDNEDRLEVALENNEFVEVVIEGDAMSPDFIPSQPEGVYLYSKYREPEKYIELDGDRLTTEDLVNLGKGRYKIKLTPTAEKRVQKSREVIDSIIKEKTVVYGITTGFGKFARTVIPINKLQELQVNLVRSHSSGVGKPLSPERCRMLLALRINVLAKGYSGISLETLKQVIEMFNASCLPYVPEKGTVGASGDLAPLSHLALGLVGEGKMWSPKSGWADAKYVLEAHGLKPVILKPKEGLALINGTQMITSLGCEAVERASAIARQADIVAALTLEVLKGTTKAFDTDIHALRPHRGQIEVAFRFRSLLDSDHHPSEIAESHRFCDRVQDAYTLRCCPQVHGVVNDTIAFVKNIITTELNSATDNPMVFANRGETISGGNFHGEYPAKALDYLAIGIHELAAISERRIERLCNPSLSELPAFLVAEGGLNSGFMIAHCTAAALVSENKALCHPSSVDSLSTSAATEDHVSMGGWAARKALRVIEHVEQVLAIELLAACQGIEFLRPLKTTTPLEKVYDLVRSVVRPWIKDRFMAPDIEAAHRLLLEQKVWEVAAPYIEKYRMEHIPESRPLSPTAFSLQFLHKKSTKIPESEDL
Specifications

Specifications

histidine ammonia-lyase
Polyclonal
Rabbit polyclonal antibody raised against a full-length human HAL protein.
HAL
HIS/HSTD
Rabbit
Affinity Purified
RUO
3034
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG
Western Blot
Unconjugated
PBS with no preservative; pH 7.4
BC096097.3
HAL
HAL (AAH96097.1, 1 a.a. ∼ 657 a.a) full-length human protein.
100 μg
Primary
Human, Mouse
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers