missing translation for 'onlineSavingsMsg'
Learn More

histidine ammonia-lyase, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human HAL protein.

Brand:  Abnova H00003034-B01.50uL

Product Code. 16198637

  • £283.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Histidine ammonia-lyase is a cytosolic enzyme catalyzing the first reaction in histidine catabolism, the nonoxidative deamination of L-histidine to trans-urocanic acid. Histidine ammonia-lyase defects cause histidinemia which is characterized by increased histidine and histamine and decreased urocanic acid in body fluids [provided by RefSeq

Sequence: MPRYTVHVRGEWLAVPCQDAQLTVGWLGREAVRRYIKNKPDNGGFTSVDDAHFLVRRCKGLGLLDNEDRLEVALENNEFVEVVIEGDAMSPDFIPSQPEGVYLYSKYREPEKYIELDGDRLTTEDLVNLGKGRYKIKLTPTAEKRVQKSREVIDSIIKEKTVVYGITTGFGKFARTVIPINKLQELQVNLVRSHSSGVGKPLSPERCRMLLALRINVLAKGYSGISLETLKQVIEMFNASCLPYVPEKGTVGASGDLAPLSHLALGLVGEGKMWSPKSGWADAKYVLEAHGLKPVILKPKEGLALINGTQMITSLGCEAVERASAIARQADIVAALTLEVLKGTTKAFDTDIHALRPHRGQIEVAFRFRSLLDSDHHPSEIAESHRFCDRVQDAYTLRCCPQVHGVVNDTIAFVKNIITTELNSATDNPMVFANRGETISGGNFHGEYPAKALDYLAIGIHELAAISERRIERLCNPSLSELPAFLVAEGGLNSGFMIAHCTAAALVSENKALCHPSSVDSLSTSAATEDHVSMGGWAARKALRVIEHVEQVLAIELLAACQGIEFLRPLKTTTPLEKVYDLVRSVVRPWIKDRFMAPDIEAAHRLLLEQKVWEVAAPYIEKYRMEHIPESRPLSPTAFSLQFLHKKSTKIPESEDL
Specifications

Specifications

histidine ammonia-lyase
Polyclonal
Mouse polyclonal antibody raised against a full-length human HAL protein.
HAL
HIS/HSTD
Mouse
50 μL
Yes
3034
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Western Blot
Unconjugated
No additive
BC096097.3
HAL
HAL (AAH96097.1, 1 a.a. ∼ 657 a.a) full-length human protein.
RUO
Primary
Human
Serum
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers