Learn More
anti-His, Monoclonal, Cytiva
Optimized for the immunodetection of histidine-tagged recombinant proteins in Western blots and ELISAs
Brand: Life Science products Cytiva 27-4710-01
Code : GE
Additional Details : Weight : 0.02000kg
Description
- Produced from a monoclonal cell line cultivated in mice (Host) and provided as clarified ascites fluid
- Provides sufficient antibody (monoclonal) for 25 Western blots
- IgG2 subclass of monoclonal antibody directed against six histidine residues, which are commonly used as an affinity tag for recombinant fusion proteins
- Offers the flexibility of detection with a secondary antibody conjugated with either horseradish peroxidase or alkaline phosphatase
- Provides high sensitivity with low background
- Volume 170μL
ELISA, Western Blotting
Specifications
Unconjugated | |
170 μL | |
Ascites |
Mouse | |
Primary | |
IgG2 |