missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-HMGB1 Ab-1 Matched Antibody Pair, Abnova™
Brand: Abnova H00003146-AP41.1Set
Additional Details : Weight : 0.00010kg
Includes: Capture antibody: mouse monoclonal anti-HMGB1, IgG2a Kappa (100μg); Detection antibody: biotinylated mouse monoclonal anti-HMGB1, IgG1 Kappa (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
Host Species: Mouse, Mouse
Isotype: IgG2a, κ, IgG1, κ
Cross Reactivity: Human
ELISA Pair
Specifications
HMGB1 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human HMGB1. | |
DKFZp686A04236/HMG1/HMG3/SBP-1 | |
Mouse | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-HMGB1, IgG2a Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-HMGB1, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Biotin | |
HMGB1 | |
HMGB1 | |
1 Set | |
3146 | |
Human | |
IgG1, IgG2a |