missing translation for 'onlineSavingsMsg'
Learn More

anti-HSPB1, MAPKAPK3 Protein Protein Interaction Antibody Pair, Abnova™

Brand:  Abnova DI0339.1Set

Product Code. 16171284

  • £549.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: HSPB1 rabbit purified polyclonal antibody (20μg); MAPKAPK3 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
This item is not returnable. View return policy

Description

Description

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the HSPB1 protein, and the other against the MAPKAPK3 protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Specifications

HSPB1
Monoclonal/Polyclonal
HSPB1
HSPB1
1 Set
Primary
Antibody pair set content: 1. HSPB1 rabbit purified polyclonal antibody (20 ug) 2. MAPKAPK3 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Purified
In situ PLA
Unconjugated
CMT2F/DKFZp586P1322/HMN2B/HS.76067/HSP27/HSP28/Hsp25/SRP27
Mouse, Rabbit
RUO
3315/7867
Human
HSPB1 & MAPKAPK3 Protein Protein Interaction Antibody Pair
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers