missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biosciences™ Anti-Human IFN-γ FITC/CD69 PE/CD4 PerCP-Cy™5.5/CD3 APC (RUO/GMP)
Monoclonal antibodies for detecting human antigens.
Brand: BD Biosciences™ 337184
This item is not returnable.
View return policy
Description
- Clone SK7 (also known as Leu-4) 25723.11 L78 SK3 (also known as Leu3a) (Ruo (GMP)).
- Anti-Hu–IFN-γ, clone 25723.11, is derived from the hybridization of mouse P3X-63- Ag8.653 myeloma cells with lymph node cells from BALB/c mice immunized with recombinant human IFN-γ.
- CD69, clone L78, is derived from hybridization of mouse Sp2/0-Ag14 myeloma cells with lymph node cells from BALB/c mice immunized with a CD8+ alloantigen-directed cytotoxic T-lymphocyte (CTL) cell line.
- CD4, clone SK3, is derived from hybridization of mouse NS-1 myeloma cells with spleen cells from BALB/c mice immununized with human peripheral blood T lymphocytes.
- CD3, clone SK7, is derived from hybridization of mouse NS-1 myeloma cells with spleen cells from BALB/c mice immunized with human thymocytes.
- Anti-Human Interferon- γ (Anti-Hu–IFN-γ) recognizes a 20- to 25-kilodalton (kd) glycoprotein.
- CD69 recognizes a very early human activation antigen. The CD69 antigen is a surface homodimer formed by the association of 28- and 32-kd chains that are held together by disulfide bridges.
- The CD4 antigen, Mr 55 kd, is present on T-helper/inducer lymphocytes and monocytes.
- CD3 reacts with the epsilon chain of the CD3 antigen/T-cell antigen receptor (TCR) complex. This complex is composed of at least six proteins that range in molecular weight from 20 to 30 kd.
- The antigen recognized by CD3 antibodies is noncovalently associated with either α/β or γ/δ TCR (70 to 90 kd).
- Contents:
- Specificity: CD3, Clone: SK7 (also known as Leu4), Format: APC, Isotype: IgG1K, Entrez Gen ID: 916
- Specificity: IFN-γ, Clone: 25723.11, Format: FITC,Isotype: IgG2bK, Entrez Gen ID: 3458
- Specificity: CD69, Clone: L78, Format: PE, Isotype: IgG1K, Entrez Gen ID: 969
- Specificity: CD4, Clone: SK3 (also known as Leu-3a), Format: PerCP-CY™5.5, Isotype: IgG1K, Entrez Gen ID: 920
- Research applications include the following:
- Studies of T-cell responses to antigens such as cytomegalovirus (CMV), human immunodeficiency virus (HIV), herpes viruses, and tumor antigens.

Specifications
CD3, IFN-γ, CD69, CD4 | |
SK7, 25723.11, L78, SK3 | |
50 Tests | |
Storage buffer: Phosphate buffered saline with BSA, beta-lactoglobulin, and 0.1% sodium azide. |
Flow Cytometry | |
Mouse | |
RUO | |
Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Biosciences™ Anti-Human IFN-γ FITC/CD69 PE/CD4 PerCP-Cy™5.5/CD3 APC (RUO/GMP) > 50 Tests
Spot an opportunity for improvement?Share a Content Correction