missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biosciences™ Anti-Human T-Lymphocyte Value Bundle 1 (RUO/GMP)
Monoclonal antibodies for detecting human antigens.
Brand: BD Biosciences™ 340407
This item is not returnable.
View return policy
Description
- Contents:
- CD4, CD69, CD3 antibody cocktail
- CD2/CD2R activation control
- CD8, CD69, CD3 antibody cocktail
- CD3 Isotype: control cocktail
- Research applications include studies of the following:
- Activated T lymphocytes and T-lymphocyte subsets (helper/suppressor) in peripheral blood.
- T-lymphocyte activation mechanisms.

Specifications
CD4, CD2R, CD69, CD3, CD8 | |
Mouse | |
RUO | |
Human |
Flow Cytometry | |
50 Tests | |
Storage buffer: Phosphate buffered saline with BSA, beta-lactoglobulin, and 0.1% sodium azide. | |
Store vials at 2°C–8°C. Conjugated forms should not be frozen. Protect from exposure to light. Each reagent is stable until the expiration date shown on the bottle label when stored as directed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Biosciences™ Anti-Human T-Lymphocyte Value Bundle 1 (RUO/GMP) > 50 Tests
Spot an opportunity for improvement?Share a Content Correction