missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ID2 Monoclonal Antibody (4E12G5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517095
This item is not returnable.
View return policy
Description
MA5-17095 targets ID2 in indirect ELISA, FACS, IHC, and WB applications and shows reactivity with Human samples. The MA5-17095 immunogen is purified recombinant fragment of human ID2 (amino acids: 1-134) expressed in E. Coli. MA5-17095 detects ID2 which has a predicted molecular weight of approximately 15kDa.
The inhibitor of DNA binding 2 (ID2) protein is essential for promoting the development of a number of cell types including natural killer cell, erythroid precursors, as well as, CD8+ and CD4+ T-cell. A lack of ID2 promotes B-cell development. The chief role of this protein is to associate with ubiquitously-expressed E proteins, and other proteins of the basic helix-loop-helix (HLH) family, preventing them from binding to DNA and activating transcription. Activation of ID2 is critical for the development of the natural killer cell lineage.Specifications
ID2 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q02363 | |
ID2 | |
Purified recombinant fragment of human ID2 (amino acids: 1-134) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
4E12G5 | |
Unconjugated | |
ID2 | |
Ac2-300; AI255428; bHLHb26; C78922; cell growth-inhibiting gene 8; class B basic helix-loop-helix protein 26; DNA-binding protein inhibitor ID2; DNA-binding protein inhibitor ID-2; GIG8; helix-loop-helix protein ID2; Id2; Id-2; ID2A; ID2H; Idb2; Inhibitor of differentiation 2; inhibitor of DNA binding 2; inhibitor of DNA binding 2, dominant negative helix-loop-helix protein; inhibitor of DNA binding 2, HLH protein; MGC26389; OTTHUMP00000140258; OTTHUMP00000200297 | |
Mouse | |
Protein G | |
RUO | |
3398 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |