missing translation for 'onlineSavingsMsg'
Learn More

anti-IGF1R, STAT3 Protein Protein Interaction Antibody Pair, Abnova™

Brand:  Abnova DI0481.1Set

Additional Details : Weight : 0.00010kg

Product Code. 16141404

  • £529.00 / 1 set
Estimated Shipment: 06-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: IGF1R rabbit purified polyclonal antibody (20μg); STAT3 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols

Description

Description

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the IGF1R protein, and the other against the STAT3 protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Specifications

IGF1R
Monoclonal/Polyclonal
IGF1R
IGF1R
1 Set
Primary
Antibody pair set content: 1. IGF1R rabbit purified polyclonal antibody (20 ug) 2. STAT3 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Purified
In situ PLA
Unconjugated
CD221/IGFIR/JTK13/MGC142170/MGC142172/MGC18216
Mouse, Rabbit
RUO
3480/6774
Human
IGF1R & STAT3 Protein Protein Interaction Antibody Pair
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers