missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IgG, BUV496; Clone: A19-3; BD
Brand: BD Biosciences 564660
This item is not returnable.
View return policy
Description
Applications:
Flow cytometry, Isotype control

Specifications
IgG | |
A19-3 | |
Brilliant Ultraviolet 496 | |
TNP-keyhole limpet hemocyanin | |
RUO | |
Store undiluted at 4deg.C and protected from prolonged exposure to light. Do not freeze |
Flow Cytometry | |
0.2mg/mL | |
Hamster | |
Affinity Purified | |
Secondary | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
anti-IgG, BUV496; Clone: A19-3; BD > 50μg; BUV496
Spot an opportunity for improvement?Share a Content Correction