Learn More
IgG (H+L) Sheep anti-Human, FITC, Invitrogen™
Sheep Polyclonal Secondary Antibody
Brand: Invitrogen PA516924
Code : A0
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
PA5-16924 targets IgG in IF applications and shows reactivity with human gamma-chain IgG. The PA5-16924 immunogen is IgG from human serum.
Specifications
Human IgG (H+L) | |
Polyclonal | |
TBS with 1% carrier protein and 0.1% sodium azide; pH 7.4 | |
IgG from human serum | |
2 mL | |
Secondary | |
4° C | |
IgG |
Immunofluorescence | |
FITC | |
Sheep | |
Purified | |
RUO | |
Human | |
Liquid |