missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG1 κ, Mouse, FITC, Clone: MOPC-21, Isotype Control, BD

Brand:  BD Biosciences 554679

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10014743

  • £313.00 / Each
Estimated Shipment: 08-08-2024
to see stock.
Add to basket

Description

Description

The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1 κ
Species Reactivity: Non-human primate

Flow Cytometry, Intracellular Staining, Isotype Control

Specifications

Specifications

Flow Cytometry, Western Blot
0.5mg/mL
Mouse
0.1 mg
Undiluted at 4°C
MOPC-21
FITC
Affinity Purified
Non-human Primate
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers