Learn More
anti-IgG1 κ, Mouse, PerCP-Cy™5.5, Clone: MOPC-21, Isotype Control, BD
For low background screening on a variety of mouse and human tissues. anti-IgG1 κ, Mouse, PerCP-Cy™5.5, Clone: MOPC-21, Isotype Control, BD immunoglobulin is a mouse myeloma protein. This antibody is routinely tested by flow cytometric analysis.
Brand: BD Biosciences 552834
Additional Details : Weight : 0.01000kg
Description
This reagent has been used as isotype control in immunophenotypic studies with baboon, rhesus, or cynomolgus peripheral blood cells. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
- Host Species: Mouse
- Clone: MOPC-21
- Isotype: IgG1 κ
- Applications: Flow Cytometry, Isotype Control
Specifications
Flow Cytometry | |
MOPC-21 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
IgG1 κ |
Monoclonal | |
PerCP-Cyanine5.5 | |
Mouse | |
50 Tests | |
Undiluted at 4°C. Protect from exposure to light. Do not freeze. |