Learn More
anti-IgG2a κ, Mouse, APC, Clone: G155-178, Isotype Control, BD
£387.00 - £541.00
Specifications
Clone | G155-178 |
---|---|
Applications | Flow Cytometry |
Conjugate | APC |
Host Species | Mouse |
Target Species | Non-human Primate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15866418
|
BD Biosciences
550882 |
0.1 mg |
£541.00
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
15846648
|
BD Biosciences
551414 |
50 Tests |
£387.00
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
Description
The G155-178 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. The immunoglobulin from clone G155-178 was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: G155-178
Isotype: IgG2a κ
Species Reactivity: Non-human primate
Immunogen: TNP-keyhole limpet hemocyanin
Flow Cytometry, Isotype Control
Specifications
G155-178 | |
APC | |
Non-human Primate | |
IgG2a κ | |
Undiluted at 4°C. Protect from exposure to light. Do not freeze. |
Flow Cytometry | |
Mouse | |
TNP-keyhole limpet hemocyanin | |
Affinity Purified |