missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG2a, PE, Clone: X39, BD

Brand:  BD Biosciences 340459

Additional Details : Weight : 0.03000kg

Product Code. 15855888

  • £319.00 / Each
Estimated Shipment: 12-07-2024
to see stock.
Add to basket

Description

Description

Flow Cytometry

Specifications

Specifications

Flow Cytometry
PE
Affinity Purified
Isotype Control (Primary)
IgG2a κ
X39
Mouse
25 μg
Undiluted at 2° to 8°C. Protect from exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers