missing translation for 'onlineSavingsMsg'
Learn More

anti- IgG2b κ, Mouse, Alexa Fluor™ 647, Clone: 27-35, BD

Brand:  BD Biosciences 557903

Additional Details : Weight : 0.30000kg

Product Code. 15633194

  • £397.00 / Each
Estimated Shipment: 08-08-2024
to see stock.
Add to basket

Description

Description

The mouse IgG2b, κ immunoglobulin isotype control monoclonal antibody 27-35 is specific for the hapten dansyl (5-[dimethylamino] naphthalene-1-sulfonyl). This hapten is not expressed on human cells or human cell lines. The 27-35 immunoglobulin was selected as an isotype control following testing which demonstrated low background staining on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: 27-35
Isotype: IgG2b, κ

Flow Cytometry, Isotype Control

Specifications

Specifications

Flow Cytometry, Immunocytology
Brilliant Violet 421
TNP-keyhole limpet hemocyanin
50 μg
IgG2a κ
G155-178
Mouse
Affinity Purified
4°C, protect from prolonged exposure to light, DO NOT FREEZE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers