missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Serological Pipettes, 2 ml, graduated 1/100 ml
GREENER_CHOICE

High optical clarity, maximum accuracy

Brand:  Greiner Bio-One 710107

Product Code. 10480803

  • £336.00 / Pack of 1000

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Free of detectable DNase, RNase, human DNA  
  • Non-Pyrogenic, non-cytotoxic
  • Sterile
  • High optical clarity
  • Maximum accuracy
  • Drop-free pipetting
  • Clear, easily legible graduation
  • Pipetting units available on request
  • Pipette colour code according to international standards
  • Maximum clarity from high-grade polystyrene
  • Lot number and best-before date on each bag
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Shape: Standard
Yes
25 pieces/bag
Clear
Serological pipets
Sterile
1/100
Bulk
2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers