Learn More
IL-6 Rat anti-Human, Biotin, Clone: MQ2-39C3, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 554546
Additional Details : Weight : 0.01000kg
Description
The MQ2-39C3 antibody reacts with human interleukin-6 (IL-6). The immunogen used to generate the MQ2-39C3 hybridoma was recombinant human IL-6. This is a neutralizing antibody.
Host Species: Rat
Clone: MQ2-39C3
Isotype: IgG2a
Species Reactivity: Human
Immunogen: Recombinant human IL-6
ELISA, Western Blotting
Specifications
IL-6 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant human IL-6 | |
0.5 mg | |
Immunology | |
Human | |
IgG2a |
ELISA | |
MQ2-39C3 | |
Biotin | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.