Learn More
IL-6 Rat anti-Human, NA/LE, Unlabeled, Clone: MQ2-13A5, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 554541
Additional Details : Weight : 0.01000kg
Description
The MQ2-13A5 antibody reacts with human interleukin-6 (IL-6). The immunogen used to generate this hybridoma was COS-7 -expressed recombinant human IL-6. This is a neutralizing antibody.
Specifications
IL-6 | |
Monoclonal | |
1.0mg/mL | |
IL6; Interleukin-6; BSF-2; CDF; HGF; HSF; IFNB2 | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
ELISA | |
MQ2-13A5 | |
Unconjugated | |
No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay. | |
Human IL-6 Recombinant Protein | |
0.5 mg | |
Immunology | |
Human | |
IgG1 |
For Research Use Only.