Learn More
anti-IL6, HRH1 Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0229.1Set
Additional Details : Weight : 0.02000kg
Includes: IL6 rabbit purified polyclonal antibody (20μg); HRH1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the IL6 protein, and the other against the HRH1 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
IL6 | |
Monoclonal/Polyclonal | |
IL6 | |
IL6 | |
1 Set | |
Primary | |
Antibody pair set content: 1. IL6 rabbit purified polyclonal antibody (20 ug) 2. HRH1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
BSF2/HGF/HSF/IFNB2/IL-6 | |
Mouse, Rabbit | |
RUO | |
3569/3269 | |
Human | |
IL6 & HRH1 Protein Protein Interaction Antibody Pair |