missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-ITGB5 Matched Antibody Pair, Abnova™
Brand: Abnova H00003693-AP51.1Set
Additional Details : Weight : 0.00010kg
Includes: Capture antibody: mouse monoclonal anti-ITGB5 (100μg); Detection antibody: rabbit purified polyclonal anti-ITGB5 (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ITGB5
Host Species: Mouse, Rabbit
Species Reactivity: Hu
ELISA Pair (Transfected Lysate)
Specifications
ITGB5 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ITGB5. | |
FLJ26658 | |
Mouse, Rabbit | |
RUO | |
3693 | |
Human | |
Purified |
ELISA | |
Unconjugated | |
ITGB5 | |
ITGB5 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-ITGB5 (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-ITGB5 (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |