Learn More
anti-KHK Matched Antibody Pair, Abnova
Brand:  Abnova H00003795-AP11.1Set
Additional Details : Weight : 0.00010kg
Product Code. 16036428
  • £660.00 / 1 set
Estimated Shipment: 02-08-2024
to see stock.
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-KHK (100μg); Mouse monoclonal anti-KHK, IgG2aκ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
Description
This gene encodes ketohexokinase that catalyzes conversion of fructose to fructose-1-phosphate. The product of this gene is the first enzyme with a specialized pathway that catabolizes dietary fructose. Alternatively spliced transcript variants encoding different isoforms have been identified.
ELISA
Specifications
Specifications
KHK
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human KHK.
KHK
Affinity Purified
RUO
3795
Human
IgG2a
ELISA
Unconjugated
KHK
Mouse, Rabbit
1 Set
Primary
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-KHK (100 μg) 2. Detection antibody: mouse monoclonal anti-KHK, IgG2a Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.