Learn More
LNX2 Recombinant Polyclonal Antibody (22HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Brand: Invitrogen 710665
Description
This antibody is predicted to react with Goat, Bovine, Horse, Pig, Cat and Rabbit. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
LNX2 Recombinant Polyclonal antibody specifically detects LNX2 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
LNX2 | |
Recombinant Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q8N448 | |
LNX2 | |
3 synthetic peptides corresponding to human LNX2 (aa 129-145, 442-458, 671-683) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
22HCLC | |
Unconjugated | |
LNX2 | |
9630046H24; AW209022; ligand of Numb protein X 2; ligand of numb-protein X 2; Lnx2; Numb-binding protein 2; PDZ domain containing ring finger 1; PDZ domain-containing RING finger protein 1; PDZRN1 | |
Rabbit | |
Protein A | |
RUO | |
222484 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |