Learn More
Ambion™ Anti-miR™ miRNA Inhibitor
Product Code. p-7075634
Click to view available options
Quantity:
20 nmol
5 nmol
Unit Size:
Each
For Research Use Only. All usage must comply with product instructions.
Product Code. 10299484
Brand: Ambion™ AM17000
For Research Use Only. All usage must comply with product instructions.
Anti-miR™ miRNA Inhibitor
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage The Anti-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. It is guaranteed for 1 year when stored dried at –20°C. Keep in a dry, cool and well-ventilated place
Description Anti-miR™ miRNA Inhibitor, 5nmol, Solid, Powder, Keep in a Dry, Cool and Well-Ventilated Place
Final Product Type Anti-miR™ miRNA Inhibitor
Packaging Type 1 Each
Product Line Ambion, Anti-miR
Product Type Anti-miR™ miRNA Inhibitor
Quantity 5 nmol
Sample Size 5nmol
Form Powder