missing translation for 'onlineSavingsMsg'
Learn More

F-Actin Mouse anti-Human, Mouse, Rabbit, Rat, Clone: NH3, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 6 publications

Brand:  Novus Biologicals NB100-64792

Additional Details : Weight : 0.00970kg

Product Code. 15449893

  • £519.00 / 0.25mg
Estimated Shipment: 11-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Actin Gamma 1 Monoclonal specifically detects Actin Gamma 1 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

F-Actin
Monoclonal
1.0 mg/mL
Western Blot 1:100-1:500, Flow Cytometry 1:10, ELISA 1:10, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
P63261
ACTG1
Human monocytes and U937 cell line
0.25 mg
Primary
Recognizes human Filamentous actin (F-actin). The binds to the N-terminal region of actin, but not to the extreme N-terminal 40 amino acids. In tissue sections the stains the cytoplasm of macrophages strongly, and gives granular, localized nuclear staining of all cell types.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgM
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
NH3
Unconjugated
Phosphate buffered saline with 0.09% Sodium Azide
actin, cytoplasmic 2, DFNA26, ACT, ACTG, actin, gamma 1, BRWS2, DFNA20
Mouse
IgM purified
RUO
71
Human, Mouse, Rat, Rabbit
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only