Learn More
anti-Mouse IgG, Biotinylated Species-specific Whole Antibody (from sheep), Cytiva
Brand: Life Science products Cytiva RPN1001-2ML
Code : GE
Additional Details : CAS Number : 26628-22-8 Weight : 0.60330kg
Description
Host Species: Sheep
Isotype: IgG
Species Reactivity: Ms
Immunodetection, ELISA, In situ Hybridization, Protein Blotting, ELISA, Immunocytochemistry
Specifications
IgG, Whole Molecule | |
Biotin | |
Affinity Purified | |
Primary |
Monoclonal | |
Sheep | |
2 mL | |
IgG |