missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Mouse IgG, peroxidase-linked species-specific whole antibody (from sheep) Secondary Antibody, Cytiva
Highly species-specific anti-Mouse IgG. Cytiva Lifescience™ anti-Mouse IgG, peroxidase-linked species-specific whole antibody (from sheep) Secondary Antibody is designed for applications such as protein blotting, ELISA and immunocytochemistry.
Brand: Life Science products Cytiva NA931-100UL
This item is not returnable.
View return policy
Description
Host Species: Sheep
Isotype: IgG
Species Reactivity: Ms
Specifications
IgG, Whole Molecule | |
HRP | |
Affinity Purified | |
Secondary |
Monoclonal | |
Sheep | |
0.1 mL | |
IgG |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains :
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
anti-Mouse IgG, peroxidase-linked species-specific whole antibody (from sheep) Secondary Antibody, Cytiva > 0.1mL; HRP
Spot an opportunity for improvement?Share a Content Correction