missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgG1 Isotype Control (CT6), Invitrogen™
Mouse Isotype Control
Brand: Invitrogen SA135641
This item is not returnable.
View return policy
Description
SA1-35641 is a mouse IgG1 negative control antibody. This antibody may be used for the detection of guinea pig lymphocyte subset, and as a negative control for mouse monoclonal antibodies of the same IgG subclass. It does not cross react with the 170-180 kDa glycoprotein which is strongly correlated with the degree of Multi-Drug-Resistance.
Mouse IgG1 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG1 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG1 is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.Specifications
Mouse IgG1 | |
CT6 | |
Assay Dependent | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
proprietary | |
1 mL | |
Isotype Control (Primary) | |
4° C | |
IgG1 |
Control, Western Blot | |
Unconjugated | |
PBS with 0.1% sodium azide | |
Mouse | |
purified | |
RUO | |
Not Applicable | |
Liquid |