missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG2a, Pacific Blue™, Isotype Control, Invitrogen™

Mouse Isotype Control

Brand:  Invitrogen MG2A28

Product Code. 10353833

  • £118.00 / Each
Estimated Shipment: 25-11-2024
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mouse IgG2a
100 ug/ml
PBS with 4mg/mL BSA and 0.1% sodium azide
Mouse
Purified
RUO
None
Liquid
Flow Cytometry
Pacific Blue
IgG2, IgG2a
Mouse IgG2a
500 μL
Isotype Control (Primary)
Store at 4°C, in the dark
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers