Learn More
NKB1 Mouse anti-Human, Unlabeled, Clone: DX9, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555964
Additional Details : Weight : 0.01000kg
Description
Reacts with the killer cell inhibitory receptor (KIR), NKB1, a 70kDa glycoprotein, member of the Ig superfamily, expressed on a subset of natural killer cells and a small subset of T cells. Expression of NKB1 has been observed to vary among individuals. KIR molecules specifically recognize a certain group of HLA class I antigens. Interaction of NKB1 with specific HLA-B antigen on a target cell inhibits cell mediated cytotxicity, possibly by delivering a negative signal preventing lymphocyte activation. It is suggested that this MHC class I-KIR interaction works as a regulatory mechanism of NK and T-cell responses to antigenic challenge. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: DX6
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
NKB1 | |
Monoclonal | |
0.5mg/mL | |
KIR3DL1; CD158E; CD158E1; KIR antigen 3DL1; KIR; KI3L1; NKAT-3; NKB1; NKB1B | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
DX9 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human NK Cell Clone | |
0.1 mg | |
Immunology | |
Human | |
IgG1 κ |
For Research Use Only.