Learn More
anti-NNMT Matched Antibody Pair, Abnova
Brand: Abnova H00004837-AP11.1Set
Additional Details : Weight : 0.00010kg
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-NNMT (100μg); Mouse monoclonal anti-NNMT, IgG1κ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
N-methylation is one method by which drug and other xenobiotic compounds are metabolized by the liver. This gene encodes the protein responsible for this enzymatic activity which uses S-adenosyl methionine as the methyl donor.
ELISA
Specifications
NNMT | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human NNMT. | |
NNMT | |
Affinity Purified | |
RUO | |
4837 | |
Human | |
IgG1 |
ELISA | |
Unconjugated | |
NNMT | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-NNMT (100 μg) 2. Detection antibody: mouse monoclonal anti-NNMT, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |