missing translation for 'onlineSavingsMsg'
Learn More

p16[INK4] Mouse anti-Human, Unlabeled, Clone: G175-405, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 550834

Additional Details : Weight : 0.01000kg

Product Code. 15896398

  • £299.00 / Each
Estimated Shipment: 30-08-2024
to see stock.

Description

Description

Cyclins and cyclin-dependent kinases (cdks) form active complexes that regulate key events during the progression of the cell cycle and are evolutionarily highly conserved. The p16 protein has been identified as a specific inhibitor of cdk4 because it blo

Host Species: Mouse
Clone: G175-405
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human p16[INK4a] fusion protein

Immunohistochemistry, Immunohistochemistry (Frozen)

Specifications

Specifications

p16[INK4]
Monoclonal
Unconjugated
Human p16[INK4a] fusion protein
1 mL
Cell Biology
Human
Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
G175-405
Mouse
Affinity Purified
RUO
Primary
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.