Learn More
anti-p53, Clone: Y5, Abcam™
Brand: Abcam AB32049-100UL
Code : Z1
Additional Details : Weight : 0.25000kg
Description
Host Species: Rabbit
Clone: Y5
Isotype: IgG
Cross Reactivity: Human. Does not react with Mouse, Rat
Immunogen: A synthetic peptide corresponding to N-terminal residues human p53
Applications:
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting
Specifications
p53 | |
Y5 | |
Rabbit | |
100 μL | |
Primary | |
IgG |
Monoclonal | |
Unconjugated | |
A synthetic peptide corresponding to N-terminal residues human p53 | |
Cancer and Tumor Biology, Cell Biology, Epigenetics And Nuclear Signaling | |
Human |