Learn More
anti-PDK3, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00005165-PW1.1Set
Additional Details : Weight : 0.02000kg
Product Code. 16037788
  • £660.00 / 1 set
Estimated Shipment: 02-08-2024
to see stock.
Includes: Antibody pair for IP: rabbit polyclonal anti-PDK3 (300μL), Antibody pair for WB: mouse monoclonal anti-PDK3 (50μg)
Specifications
PDK3
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
PDK3
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-PDK3 (300 ul) 2. Antibody pair for WB: mouse monoclonal anti-PDK3 (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Immunoprecipitation, Western Blot
Unconjugated
PDK3
Mouse, Rabbit
RUO
5165